Pesticide surrogate mix (2 components)
Catalog # REST32453
Supplier: Restek
CAS Number:
Specifications
- Pk:1 mL
- Description:Pesticide surrogate mix, in acetone
- Environmentally Preferable:
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains two components, those are decachlorobiphenyl and 2,4,5,6-tetrachloro-m-xylene.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.