Order Entry
Slovenia
ContactUsLinkComponent
TX TPH calibration mix (2 components)
TX TPH calibration mix (2 components)
Catalog # REST31483
Supplier:  Restek
CAS Number:  
undefined
TX TPH calibration mix (2 components)
Catalog # REST31483
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    10000 μg/ml each in pentane
  • Pk:
    1 mL
  • Description:
    TX TPH calibration mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The mix consists of diesel fuel #2 composite, unleaded gasoline composite.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.