JP-5 military fuel standard
Catalog # REST31252
Supplier: Restek
CAS Number:
Specifications
- Concentration:50000 μg/ml in methylene chloride
- Pk:1 mL
- CAS No.:8008-20-6
- Description:JP-5 military fuel
- Environmentally Preferable:
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.