Order Entry
Slovenia
ContactUsLinkComponent
 
MA EPH surrogate spike mix (2 components)
Catalog # REST31479
Supplier:  Restek
CAS Number:  
undefined
MA EPH surrogate spike mix (2 components)
Catalog # REST31479
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    4000 µg/ml each in acetone
  • Pk:
    1 mL
  • Description:
    MA EPH surrogate spike mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

This spike mix contains two components: 1-Chlorooctadecane and o-Terphenyl.

Shelf life 6 months.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.