Pesticide mix #1 (rev), organophosphorus, method 525.2 (7 components)
Catalog # REST33013
Supplier: Restek
CAS Number:
Specifications
- Concentration:500 µg/ml each in acetone
- Pk:1 mL
- Description:Pesticide mix #1, organophosphorus
- Pk Info:1 ml/ampoule
- Type:Pesticide mixture
Specifications
About this item
Contains: Chlorpyrifos, dichlorvos (DDVP), disulfoton sulfone, ethoprop (ethoprophos), methyl paraoxon (parathion methyl-o-analog), mevinphos, tetrachlorvinphos.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.