Pesticide mix #1 (rev), organonitrogen, method 525.2 (37 components)
Catalog # REST33012
Supplier: Restek
CAS Number:
Specifications
- Concentration:500 µg/ml each in acetone
- Pk:1 Ampoul
- Description:Pesticide mix #1, organonitrogen
- Pk Info:1 ml/ampoule
- Type:Pesticide mixture
Specifications
About this item
Contains: Alachlor, ametryn, atraton, atrazine, bromacil, butachlor, butylate, chlorpropham, cyanazine (bladex), cycloate, diphenamid, EPTC, etridiazole, fenarimol, fluridone (sonar), hexazinone (velpar), metolachlor, metribuzin, MGK-264, molinate, napropamide (devrinol), norflurazon, pebulate, prometon, prometryne, propachlor, propazine, propyzamide, simazine, simetryn, tebuthiuron, terbacil, terbutryn, triadimefon, tricyclazole (beam), trifluralin, vernolate.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.