524 calibration mix #7B (7 components)
REST30304
: Restek
:
- Concentration:2000 µg/ml each in P and T methanol
- Pk:1 mL
- Description:524 Calibration Mix #7B
- Pk Info:1 ml/ampoule
The calibration mix consists of seven components they are acrylonitrile, allyl chloride (3-chloropropene), ethyl methacrylate, methyl acrylate, methyl methacrylate, nitrobenzene and pentachloroethane.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.