MA VPH surrogate standard (2,5-dibromotoluene)
Catalog # REST30453
Supplier: Restek
CAS Number:
Specifications
- Concentration:10000 µg/ml in P and T methanol
- Pk:1
- Description:2,5-Dibromotoluene
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.