LC multiresidue pesticide standard #9 (7 components)
Catalog # REST31980
Supplier: Restek
CAS Number:
Specifications
- Concentration:100 µg/ml each in acetonitrile
- Pk:1 Ampoul
- Description:LC multiresidue pesticide standard #9
- Pk Info:1 ml/ampoule
Specifications
About this item
Accurately detect and quantify pesticides of global food safety concern in a wide range of fruits, vegetables, and other commodities by LC-MS/MS.
- Quantitatively tested to confirm composition, detailed support documentation provided
- Carbamate/uron compounds
Formulated and grouped for maximum long-term stability* and well-balanced chromatographic performance.
Optimised multiresidue pesticide method is offered free of charge and downloadable XLS file includes conditions and transition tables.
Consists of 7 components, those are aminocarb, desmedipham, formetanate hcl, mexacarbate, monceren, phenmedipham and propamocarb free base.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Ordering information: The complete 204-compound kit also available (cat. # 31971) that covers many LC-determined pesticides listed by government agencies, other ampoules also sold separately.
Caution: * NOTE: When combining many compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 204 compounds involved, and then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analysing each mix separately to ensure accurate results for every compound.