8040 Surrogate mix (2 components)
Catalog # REST31090
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 μg/ml each in isopropanol
- Pk:1 mL
- Description:8040 Surrogate mix
- Pk Info:1 ml/ampoule
Specifications
About this item
8040 Surrogate mix contains 2-fluorophenol, 2,4,6-tribromophenol.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.