PCB congener standard #1
Catalog # REST32290
Supplier: Restek
CAS Number:
Specifications
- Concentration:10 µg/ml each in isooctane
- Pk:1
- Description:PCB Congener Standard #1
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains 6 components: 2,4,4'-trichlorobiphenyl (BZ #28), 2,2',5,5'-tetrachlorobiphenyl (BZ #52), 2,2',4,5,5'-pentachlorobiphenyl (BZ #101), 2,2',3,4,4',5'-hexachlorobiphenyl (BZ #138), 2,2',4,4',5,5'-hexachlorobiphenyl (BZ #153), 2,2',3,4,4',5,5'-heptachlorobiphenyl (BZ #180).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.