Order Entry
Slovenia
ContactUsLinkComponent
Pentacosane (C25)
Pentacosane (C25)
Catalog # REST31487
Supplier:  Restek
CAS Number:  
undefined
Pentacosane (C25)
Catalog # REST31487
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    10,000 μg/ml in methylene chloride
  • Pk:
    1
  • CAS No.:
    629-99-2
  • Description:
    Pentacosane
  • Formula:
    C₂₅H₅₂
  • Description:
    Pentacosane (C25)
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.