DCPA methyl ester (chlorthal-dimethyl)
Catalog # REST32216
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/mL in methanol
- Pk:1
- CAS No.:1861-32-1
- Description:DCPA methyl ester (Chlorthal-dimethyl)
- Formula:C₁₀H₆Cl₄O₄
- Description:DCPA methyl ester (Chlorthal-dimethyl)
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.