Chlorpyrifos
Catalog # REST32212
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml in methanol
- Pk:1
- CAS No.:2921-88-2
- Description:Chlorpyrifos
- Formula:C₉H₁₁Cl₃NO₃PS
- Description:Chlorpyrifos
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.