Order Entry
Slovenia
Orders LinkContactUsLinkComponent
FAME #7 mix

FAME #7 mix

FAME #7, each mixture has minimum 50 mg/ampoule, Ampoule, 1 mL
Catalog # REST35016
Supplier:   Restek
FAME #7 mix
Catalog # REST35016
Supplier:  Restek
CAS Number:  
FAME #7 mix
FAME #7 mix

FAME #7, each mixture has minimum 50 mg/ampoule, Ampoule, 1 mL

CAS Number:  
Price
109,67 €
Log in for availability
1 * 1 mL
Product Details & Documents

This mixture can be used for quantification (AOCS method CE 1a–13) and approximate the compositions of oils.

  • Suitable for oils of short chain lengths (C6 - C10)

This mix contains five components such as C6:00 methyl caproate (20%), C7:00 methyl heptanoate (20%), C8:00 methyl caprylate (20%), C9:00 methyl nonanoate (20%), C10:00 methyl caprate (20%).

Specifications

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1 mL
  • Description:
    FAME #7, each mixture has minimum 50 mg/ampoule

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.