FAME #4 mix
Catalog # REST35013
Supplier: Restek
CAS Number:
Specifications
- Pack type:Ampoule
- Pk:1
- Description:FAME #4, each mixture has minimum 50 mg/ampoule
Specifications
About this item
This mixture can be used for quantification (AOCS method CE 1a–13) and approximate the compositions of oils.
- Suitable for oils of mid-range to long chain lengths (C16 - C24)
This mix contains five components such as C16:00 methyl palmitate (20%), C18:00 methyl stearate (20%), C20:00 methyl arachidate (20%), C22:00 methyl behenate (20%), C24:00:00 methyl lignocerate (20%).