FAME #3 mix
Catalog # REST35012
Supplier: Restek
CAS Number:
Specifications
- Pack type:Ampoule
- Pk:1
- Description:FAME #3, each mixture has minimum 50 mg/ampoule
Specifications
About this item
This mixture can be used for quantification (AOCS method CE 1a–13) and approximate the compositions of oils.
- Suitable for oils of short to mid-range chain lengths (C8 - C16)
This mix contains five components such as C8:00 methyl caprylate (20%), C10:00 methyl caprate (20%), C12:00 methyl laurate (20%), C14:00 methyl myristate (20%), C16:00 methyl palmitate (20%).