Order Entry
Slovenia
ContactUsLinkComponent
Vacuum manifolds, V20, Sigma-Aldrich®
Vacuum manifolds, V20, Sigma-Aldrich®
  SIALVM20-1SET
 :  Merck
Vacuum manifolds, V20, Sigma-Aldrich®
  SIALVM20-1SET
 :  Merck
  • Description:
    VM20 Vacuum manifold set
  • Pk:
    1 SET
The VM20 vacuum manifold is designed to provide increased efficiency and performance when used with column-based nucleic acid purification systems.
  • Dual port design allows waste to be collected in the manifold body or diverted to any auxiliary reservoir
  • Stepped design for easy access to every column
  • Endura-Luer stopcocks instead of plugs
  • Chemically resistant PP
This manifold is capable of processing 20 samples simultaneously and includes a stepped design for easy access to every column. Can be used for the isolation of circulating tumor cells by filtration with track-etched membranes.
 : The kit contains the VM20 vacuum manifold, 20 Endura-Luer stopcocks, and VM20 accessory pack with cap and two nozzles.