
Poly(vinylsulphonic acid) sodium salt 30% (w/w) in water, Technical Grade, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9002-97-5
:
Formula:(C₂H₃NaO₃S)n
MDL Number:MFCD00084471
Density:1.267 g/mL (25 °C)
:M-Clarity™ quality level = MQ200
:GHS07
Specification Test Results
- Appearance (Color):Yellow to Amber
- Appearance (Form):Liquid
- Residue on Evaporation:25.0 - 35.0 %
Specification Test Results
Frequently Bought Together