Anti-IgG Goat Polyclonal Antibody (FITC)
OZBIIGG0100
- Antibody type:Control
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Goat
- Isotype:IgG
- Form:liquid
- Storage buffer:DPBS, pH 7,4
- Storage temperature:–20 °C
- Shipping temperature:RT
- Size:100 µl
- Pk:100 µl
Immunoglobulin (IgG) from goat serum has been labelled with the fluorescein isothiocyanate dye to produce a green-emitting antibody with a high fluorophore-to-protein ratio. The resulting antibody can be used as a positive control for intracellular antibody delivery with Ab-DeliverIN reagent.
Absorption maximum: 492 nm
Emission maximum: 520 nm
Extinction coefficient: 75000 M⁻¹cm⁻¹
Quantum yield: 0,79
Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Goat
Isotype: IgG
Reactivity: