Organic reference standard, sodium cetostearyl sulphate, EP reference standard
SIALS0710000
: Merck
:
- Pk:1
- Standards:Reference standard
- CAS No.:59186-41-3
- Description:Sodium cetostearyl sulphate
- Formula:C₃₄H₇₀NaO₈S₂
- Description:Sodium cetostearyl sulphate
- Molecular weight (Mw):694,04 g/mol