Order Entry
Slovenia
ContactUsLinkComponent
Simulated gastric fluid (without enzyme) concentrate for dilution to (NaCl 2 g/L), DILUT-IT® dissolution media concentrate, J.T.Baker®
Simulated gastric fluid (without enzyme) concentrate for dilution to (NaCl 2 g/L), DILUT-IT® dissolution media concentrate, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Simulated gastric fluid (without enzyme) concentrate for dilution to (NaCl 2 g/L), DILUT-IT® dissolution media concentrate, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    1.9 - 2.1 g/l
  • Assay, Sodium Chloride (NaC₁) (at 26x dilution):
    1.9 - 2.1 g/l
  • pH at 25°C at 26x Dilution:
    1.15 - 1.25

Specification Test Results

Frequently Bought Together