Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-HHV8 Rat Monoclonal Antibody [clone: LN53]
  VWRKILM3935-C1
Anti-HHV8 Rat Monoclonal Antibody [clone: LN53]
  VWRKILM3935-C1

 

  • Antibody type:
    Primary
  • Antigen symbol:
    HHV8
  • Clonality:
    Monoclonal
  • Clone:
    LN53
  • Conjugation:
    Unconjugated
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2c
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Concentrated
  • Storage temperature:
    2°C - 8°C
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Type: Primary
Antigen: HHV8
Clonality: Monoclonal
Clone: LN53
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2c
Reactivity: Human