Order Entry
Slovenia
ContactUsLinkComponent
Human Recombinant TNF-A (from E. coli)
Human Recombinant TNF-A (from E. coli)
Certificates
undefined
Human Recombinant TNF-A (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity assay: Cytolysis of mouse L929 cells in the presence of actinomycin D, ED50 ≤2 ng/ml, bioactivity ≥5,0×10⁵ units/mg
  • Gene ID:
    P01375
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    DIF|cachexin|cachectin|necrosin|TNFSF2|TNF|cytotoxin
  • UniProtKB:
    P01375
  • Protein/peptide name:
    TNF-alpha
  • Purity:
    ≥95%
  • Molecular weight:
    17,5 kDa
  • Sequence:
    MVRSSSRTPS DKPVAHVVAN PQAEGQLQWL NRRANALLAN GVELRDNQLV VPSEGLYLIY SQVLFKGQGC PSTHVLLTHT ISRIAVSYQT KVNLLSAIKS PCQRETPEGA EAKPWYEPIY LGGVFQLEKG DRLSAEINRP DYLDFAESGQ VYFGIIAL
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Related Information

Frequently Bought Together