12-Crown-4 (1,4,7,10-tetraoxacyclododecane, lithium ionophore V), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:294-93-9
:
Formula:C₈H₁₆O₄
MDL Number:MFCD00005103
:176.21 g/mol
Boiling Pt:254 °C (1013 hPa)
Melting Pt:16 °C
Density:1.11 g/cm³ (20 °C)
EINECS:206-036-5
ADR:6.1,II
:M-Clarity™ quality level = MQ100
UN:2810
Frequently Bought Together