Sorbitan monostearate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1338-41-6
:
Formula:C₂₄H₄₆O₆
MDL Number:MFCD00005366
:430.63 g/mol
Merck Index:14,8724
Melting Pt:53…57 °C
Density:1.00
Flash Pt:>149 °C (300 °F)
Storage Temperature:Ambient
EINECS:215-664-9
Related Information
Frequently Bought Together