Virginiamycin M1 (from Streptomyces virginiae) ~95% (by TLC), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:21411-53-0
:
Formula:C₂₈H₃₅N₃O₇
:525.6 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
EINECS:244-376-6
Specification Test Results
- C% anhydrous (Elemental analysis):63.00 - 64.50%
- N% anhydrous (Elemental analysis):7.70 - 8.70%
- Purity (TLC):≥93.00%
Specification Test Results
Frequently Bought Together