Mercury (II) oxide, red 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21908-53-2
:
Formula:HgO
MDL Number:MFCD00011045
:216.59 g/mol
Merck Index:14,5882
Boiling Pt:356.6 °C (1013 hPa)
Melting Pt:∼ 500 °C (dec.)
Density:11.14
Storage Temperature:Ambient
EINECS:244-654-7
UN:1641
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Related Information
Frequently Bought Together