Order Entry
Slovenia
ContactUsLinkComponent
Acenaphthenequinone
Acenaphthenequinone
Catalog # MOLEM89996922
Supplier:  Molekula
CAS Number:  
Acenaphthenequinone
Catalog # MOLEM89996922
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :182.18 g/mol
Boiling Pt:351 °C (1013 hPa)
Melting Pt:252…255 °C
Density:1,48 g/cm³ (20 °C)
EINECS:201-441-3
 :GHS07
CAS Number:82-86-0
Formula:C₁₂H₆O₂
MDL Number:MFCD00003805