4-Hydroxyindole ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 2380-94-1 Molecular Formula: C8H7NO Formula Weight: 133.15 Melting Point: 97-99° Beilstein Reference: 114905
Synonyms: Indol-4-ol, 4-Indolol, 4-Hydroxy-1H-indole, 1H-Indol-4-ol
- CAS Number:2380-94-1
- Formula:C₈H₇NO
- MDL Number:MFCD00005667
- Molecular Weight:133.15 g/mol
- Boiling Pt:100 °C (0.01 mmHg)
- Melting Pt:97…99 °C
- Storage Temperature:Ambient
- EINECS:219-177-2
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 2380-94-1 Molecular Formula: C8H7NO Formula Weight: 133.15 Melting Point: 97-99° Beilstein Reference: 114905
Synonyms: Indol-4-ol, 4-Indolol, 4-Hydroxy-1H-indole, 1H-Indol-4-ol
- CAS Number:2380-94-1
- Formula:C₈H₇NO
- MDL Number:MFCD00005667
- Molecular Weight:133.15 g/mol
- Boiling Pt:100 °C (0.01 mmHg)
- Melting Pt:97…99 °C
- Storage Temperature:Ambient
- EINECS:219-177-2
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



