7-Nitroindole
Glass bottle for solids, 1 g
Catalog # MOLE13536321-1G
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:6960-42-5
- Formula:C₈H₆N₂O₂
- MDL Number:MFCD00005683
- Molecular Weight:162.15 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



