
Ammonium chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:12125-02-9
:MFCD00143604
Formula:NH₄Cl
MDL Number:MFCD00011420
:53.49 g/mol
Merck Index:13,00510
Melting Pt:335 °C
Density:1.5256 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:235-186-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together