Order Entry
Slovenia
ContactUsLinkComponent
Ammonium chloride, BAKER, J.T.Baker®
Irritant
Ammonium chloride, BAKER, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Ammonium chloride, BAKER, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay (dry basis):
    99.5 - 100.5 %
  • Acidity or Alkalinity:
    passes test
  • Appearance of solution:
    passes test
  • Bromides and Iodides:
    passes test
  • Calcium (Ca):
    max. 200 ppm
  • Iron (Fe):
    max. 20 ppm
  • Heavy Metals (as Pb):
    max. 10 ppm
  • Identification:
    passes test
  • Loss on Drying:
    max. 0.5 %
  • Sulfated Ash:
    max. 0.1 %
  • Sulfates (as SO4):
    max. 150 ppm
  • Limit of thiocyanate (SCN):
    passes test
  • pH of 5% Solution at 25°C:
    4.6 - 6.0

Specification Test Results

Related Information

Frequently Bought Together