Tetraethylammonium tetrafluoroborate, Sigma-Aldrich®
Catalog # SIAL242144-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: Tetrylammonium tetrafluoroborate
:
:217.06 g/mol
Melting Pt:365…368 °C
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:207-055-1
:GHS05
:GHS07
CAS Number:429-06-1
Formula:C₈H₂₀BF₄N
MDL Number:MFCD00011827