Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

Diphosphorus pentoxide, Sigma-Aldrich®

Corrosive
Supplier:   Merck
Diphosphorus pentoxide, Sigma-Aldrich®
Supplier:  Merck
Certificates
Diphosphorus pentoxide, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  Phosphorus(V) oxide, di-Phosphorus pentoxide, Phosphorus pentoxide, Phosphoric anhydride
  • CAS Number:1314-56-3
  • Formula:P₂O₅
  • MDL Number:MFCD00011440
  • Molecular Weight:141.94 g/mol
  • Merck Index:13,07441
  • Melting Pt:562 °C
  • Density:2.253 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-236-1
  • UN:1807
  • ADR:8,II
  •  :M-Clarity™ quality level = MQ200
Product Family Options

Product Information

  • Pack type
    Pk Info
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 40,60 €
    40,60 €
    1 * 250 g
       40,60 €

  • Plastic bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 67,50 €
    67,50 €
    1 * 500 g
       67,50 €

  • 6×500 g
    1
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 362,00 €
    362,00 €
    1 * 1 Each
       362,00 €