

N-(3-(Trimethoxysilyl)propyl)ethylenediamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1760-24-3
:
Formula:H₂NCH₂CH₂NHCH₂CH₂CH₂Si(OCH₃)₃
MDL Number:MFCD00008173
:222.36 g/mol
Boiling Pt:261…263 °C (1013 hPa)
Melting Pt:∼ 0 °C
Density:1.01 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:136 °C
Storage Temperature:Ambient
EINECS:217-164-6
:GHS05
:GHS07
Specification Test Results
- Solubility:(20 °C) insoluble,Hydrolysis
- Molar Mass:222.36 g/mol
- Boiling Point:261 - 263 °C (1013 hPa)
- Vapor Pressure:1.5 hPa (20 °C)
- Flash Point:136 °C
- Density:1.02 g/cm3 (20 °C)
- pH:10 (10 g/l, H2O, 20 °C)
- Ignition Point:300 °C
- Hill Formula:C8H22N2O3Si
Specification Test Results
Related Information
MERC8.19172.0100_TDS_EN MERC8.19172.0500_TDS_EN ProductPage_819172
Product specification
msds_819172
SDS
Frequently Bought Together