

Silicon carbide, powder, medium 120 grit
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:409-21-2
:
Formula:SiC
MDL Number:MFCD00049531
:40.1 g/mol
Melting Pt:2700 °C
Density:3.16
EINECS:206-991-8
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together