Tin(IV) bromide, Sigma-Aldrich®
Catalog # SIAL215961-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:438.33 g/mol
Merck Index:12,08928
Boiling Pt:202 °C (1013 hPa)
Melting Pt:29.2 °C
Density:∼3.34 g/cm³ (20 °C)
EINECS:232-184-5
UN:3260
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
CAS Number:7789-67-5
Formula:Br₄Sn
MDL Number:MFCD00011240