Indium (III) oxide ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1312-43-2
:
Formula:In₂O₃
MDL Number:MFCD00011060
Merck Index:14,4952
Melting Pt:850 °C
Density:7.18
:277.64 g/mol
Storage Temperature:Ambient
EINECS:215-193-9
Related Information
Frequently Bought Together