
Calcium fluoride, Sigma-Aldrich®
Catalog # SIAL378801-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:78.07 g/mol
Merck Index:13,01669
Boiling Pt:2500 °C (1013 hPa)
Melting Pt:1418 °C
Density:3.18 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:232-188-7
:GHS07
CAS Number:7789-75-5
Formula:CaF₂
MDL Number:MFCD00010907