Calcium carbonate, anhydrous, Reagent Grade
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:471-34-1
:
Formula:CaCO₃
MDL Number:MFCD00010906
:100.09 g/mol
Merck Index:13,01658
Melting Pt:825 °C
Density:2.71 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-439-9
Specification Test Results
- Arsenic:≤0.0003%
- Fluoride:≤0.005%
- Heavy Metals:≤0.003%
- Lead:≤0.001%
- Loss on Drying:≤2.0%
- Magnesium and Alkali Salts:≤1.0%
- Purity:>98%
Specification Test Results
Frequently Bought Together