

Ammonium hydrogen difluoride 95%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1341-49-7
:
Formula:NH₄HF₂
MDL Number:MFCD00012018
:57.04 g/mol
Merck Index:13,00498
Melting Pt:125 °C
Density:0,8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-676-4
UN:1727
ADR:8,II
:GHS05
:GHS06
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:White Flakes or scales
- Water:≤0.5 % (K.F.)
- Titration with NaOH:≥94 %
- Sulfite (SO3):≤100 ppm
- Sulfate (SO4):≤2000 ppm
- Iron (Fe):≤500 ppm
- Heavy metals (as Pb):≤200 ppm
- Impurity:≤5 % (Ammonium fluoride (NH4F))
Specification Test Results
Frequently Bought Together