Sodium dihydrogen phosphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic
CAS Number:7558-80-7
:
Formula:NaH₂PO₄
MDL Number:MFCD00003527
Density:0.9996 g/cm³ (20 °C)
Molecular Weight:119.98 g/mol
Melting Pt:∼650 °C
Merck Index:12,08806
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together