

Sodium nitrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7631-99-4
:
Formula:NaNO₃
MDL Number:MFCD00011119
:84.99 g/mol
Merck Index:13,08720
Boiling Pt:380 °C (1013 hPa)
Melting Pt:308 °C
Density:2.26 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1498
ADR:5.1,III
:M-Clarity™ quality level = MQ200
EINECS:231-554-3
REACH:01-2119488221-41
:GHS03
:GHS07
Frequently Bought Together