
Sodium iodide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7681-82-5
:
Formula:NaI
MDL Number:MFCD00003532
:149.89 g/mol
Merck Index:13,08705
Boiling Pt:1300 °C (1013 hPa)
Melting Pt:662 °C
Density:3.67 g/cm³ (20 °C)
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ100
EINECS:231-679-3
:GHS09
Storage Temperature:Ambient
Frequently Bought Together