Order Entry
Slovenia
ContactUsLinkComponent
1,1,3,3-Tetramethylguanidine 99%
Flammable
Corrosive
Irritant
1,1,3,3-Tetramethylguanidine 99%
Certificates
undefined
Flammable
Corrosive
Irritant
1,1,3,3-Tetramethylguanidine 99%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4665 to 1.4695 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together