Order Entry
Slovenia
ContactUsLinkComponent
 
Fluorescein disodium salt, high purity
  0681-100G
 :  
undefined
Fluorescein disodium salt, high purity
  0681-100G
 :  

 

  • Pack type:
    Bottle, amber
  • Pk:
    100 g
  • Seq:
    3401

 

 

  • Lambda Max:
    487 – 493 nm
  • O.D.@ Lambda Max (0.0124 g/2 l 0.001M CO₃ Buffer):
    1 – 1.25

 

 

 :
 :376.28 g/mol
 :13,04185
 :>300 °C
 :1.601 g/cm³ (20 °C)
 :Ambient
 :208-253-0
 :518-47-8
 :C₂₀H₁₀Na₂O₅
 :MFCD00167039