1,4-Diazabicyclo[2.2.2]octane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: DABCO
CAS Number:280-57-9
:
Formula:C₆H₁₂N₂
MDL Number:MFCD00006689
Boiling Pt:174
Melting Pt:155…158 °C
Density:1.14 g/cm³ (20 °C)
Molecular Weight:112.17 g/mol
Flash Pt:62
Storage Temperature:Ambient
EINECS:205-999-9
UN:1325
ADR:4.1 III
Merck Index:12,09801
:M-Clarity™ quality level = MQ200
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together