Tetrachlorocyclopropene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6262-42-6
:
Formula:C₃Cl₄
Boiling Pt:130…131 °C (1013 hPa)
Density:1.45 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:228-418-0
UN:2810
ADR:6.1,II
:177.84 g/mol
Frequently Bought Together