

Cyclopentanone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:120-92-3
:
Formula:C₅H₈O
MDL Number:MFCD00001409
:84.12 g/mol
Merck Index:13,02771
Boiling Pt:130 °C (1013 hPa)
Melting Pt:–58 °C
Density:0.941 g/cm³ (20 °C)
EINECS:204-435-9
UN:2245
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:31 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together